Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ First Aid Kit and Replacement Items

Catalog No. 19048569
Change view
Click to view available options
Product Type:
Nitrile Gloves
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-048-569 Nitrile Gloves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-048-569 Supplier Honeywell™ Supplier No. 021640

missing translation for 'validMainframeMsg'

Specifications

For Use With (Application) First Aid
Length (English) 9.5 in.
Length (Metric) 24 cm
Product Type Nitrile Gloves
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.