Learn More
Honeywell™ North™ First Aid Self-Adhesive Wrap
Catalog No. 19074578
Change view
Click to view available options
Dimensions (L x W):
5 yd. x 1 in.
Catalog No. Dimensions (L x W)
19-074578 5 yd. x 1 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-074578 Supplier Honeywell™ Supplier No. 720070H5
This item is not returnable. View return policy
Self-Adherent Wraps are sold separately and are also a component of many first aid kits. Length: 5 yards. Choice of width: 2, 3 or 4 inches.
Used to secure and protect primary dressings and other devices. Also helps immobilize injuries and provide compression. Self-adherent wrap sticks to itself without the need for adhesive, pins or clips.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Self-Adherent Wrap
Dimensions (L x W) 5 yd. x 1 in.
For Use With (Application) Agriculture, chemical, construction, fire protection, food services, forestry, medical, law enforcement, military, mining, municipal services, nuclear, oil and gas, pharmaceutical, steel and metals, transportation, utilities, welding, wind energy