Learn More
Honeywell™ North™ First Aid Supplies
Catalog No. 19027277
Change view
Click to view available options
Product Type:
Pouch
Catalog No. Product Type
19-027277 Pouch
1 options
This item is not returnable. View return policy
Catalog No. 19-027277 Supplier Honeywell™ Supplier No. 751876
This item is not returnable. View return policy
Assists in safe and proper administration of CPR. Provides maximum protection of the first responder by enhancing bite block, while assisting in proper administration of CPR by helping to maintain an open airway.
  • CPR Microshield™: The positive one-way valve inside the airway enhancing bite block is designed for maximum protection of the first responder
  • CPR Microkey™: CPR Microchield in a black mini pouch with key chain
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pouch
For Use With (Application) First Aid Kit