Learn More
MSC North Fixed Shade Lens, Glass, 5-1/4 x 4-1/2 in, Shade 10
Catalog No. 5000507982
Encompass
This fixed shade lens is made of glass, offering a 5-1/4 in wide by 4-1/2 in high viewing area. With a shade level of 10 and a green lens color, it is designed for horizontal mounting in various applications.
  • Fixed shade lens design
  • Constructed from glass material
  • Measures 5-1/4 x 4-1/2 in
  • Provides shade level 10
  • Features a green lens color
  • Designed for horizontal mounting
Catalog No. 50-005-07982 Supplier MSC Supplier No. 62269329
This fixed shade lens is made of glass, offering a 5-1/4 in wide by 4-1/2 in high viewing area. With a shade level of 10 and a green lens color, it is designed for horizontal mounting in various applications.
  • Fixed shade lens design
  • Constructed from glass material
  • Measures 5-1/4 x 4-1/2 in
  • Provides shade level 10
  • Features a green lens color
  • Designed for horizontal mounting