Learn More
MSC North Gloves, Cut A4, Puncture 1, Abrasion 4, 15 Gauge, X-Large
Catalog No. 5000508059
Encompass
These gloves are designed to be cut, puncture, and abrasion-resistant, featuring a Cut A4, Puncture 1, and Abrasion 4 rating. They are 15 gauge and X-Large in size. Constructed with nylon, they feature a foam PVC-coated palm and fingertips, presented in a black on red color scheme.
  • Knit style
  • Foam PVC coating material on palm and fingertips
  • Cut, puncture, and abrasion-resistant
  • ANSI/ISEA Cut Resistance Level A4
  • Primary material is nylon
  • ANSI/ISEA Abrasion Resistance Level 4
  • Red and black color
  • X-Large glove size
  • ANSI/ISEA Puncture Resistance Level 1
  • Black coating color
  • 15 gauge
  • Nylon blend back material
  • Nylon lining material
  • Seamless lining
  • Knit cuff material
  • Dipped coating style
  • Red cuff color
  • Unisex design
Catalog No. 50-005-08059 Supplier MSC Supplier No. 57553760
These gloves are designed to be cut, puncture, and abrasion-resistant, featuring a Cut A4, Puncture 1, and Abrasion 4 rating. They are 15 gauge and X-Large in size. Constructed with nylon, they feature a foam PVC-coated palm and fingertips, presented in a black on red color scheme.
  • Knit style
  • Foam PVC coating material on palm and fingertips
  • Cut, puncture, and abrasion-resistant
  • ANSI/ISEA Cut Resistance Level A4
  • Primary material is nylon
  • ANSI/ISEA Abrasion Resistance Level 4
  • Red and black color
  • X-Large glove size
  • ANSI/ISEA Puncture Resistance Level 1
  • Black coating color
  • 15 gauge
  • Nylon blend back material
  • Nylon lining material
  • Seamless lining
  • Knit cuff material
  • Dipped coating style
  • Red cuff color
  • Unisex design