Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Grip N™ Nitrile-Coated Hot Mill Gloves

Catalog No. 191500984
Change view
Click to view available options
Men's
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-150-0984 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-0984 Supplier Honeywell™ Supplier No. 52/7456C

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit gloves are constructed of Aramid Fiber for protection from temperatures up to 400°F.

  • Seamless knit construction ensures superior comfort and reduces hand fatigue
  • Nitrile coating on both sides of the glove to enhance grip capabilities and extend life
  • Aramid Fiber shell provides cut/slash protection
  • Cotton/polyester liner increases comfort and thermal protection
  • Machine washable
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Cuff Material Cotton
Coating Material Nitrile
Color Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Aramid Fiber
Product Type Hot Mill Gloves
Size Universal
Temperature (English) Max. 400°F
For Use With (Application) Construction, Food Service, Manufacturing, and Steel/Metal Handling
Gender Men's
Glove Cuff Color Natural
Glove Cuff Style Gauntlet
Liner Material Cotton/Polyester Blend
Series 52/7456
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.