Learn More
Honeywell™ North™ Grip N™ Gloves with Aramid Fiber
Description
- Nitrile “N” coating improves grip and extends life due to better abrasion resistance
- Knit construction provides superior comfort (no seams for chafing) and less hand fatigue
- Machine washable or dry cleanable. Permits easy recycling for longer life and maximum value with minimal shrinkage
- Ambidextrous design gives wearer four sides instead of two to further enhance value, also no sorting after recycling
- Gloves also come in a variety of styles and weights to fit your needs
- Blended design offers economical alternative
- Puts Kevlar™ aramid fibers on outside for cut/slash protection and cotton/polyester on inside for worker comfort
Specifications
Specifications
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Universal |