Learn More
MSC North Half Facepiece Respirator, Silicone, Threaded, Large
Catalog No. 5000508727
Encompass
This half facepiece respirator is constructed from silicone and features a threaded connection, designed for comfortable and secure fit in a large size.
  • Dual sealing flange
  • Speech diaphragm
  • Not SAR compatible
  • Not PAPR compatible
Catalog No. 50-005-08727 Supplier MSC Supplier No. 39687512
This half facepiece respirator is constructed from silicone and features a threaded connection, designed for comfortable and secure fit in a large size.
  • Dual sealing flange
  • Speech diaphragm
  • Not SAR compatible
  • Not PAPR compatible