Learn More
MSC North Hard Hat, Class C, G & E, 4-Point Suspension, HDPE, Yellow
Catalog No. 5000509045
Encompass
This industrial hard hat provides certified head protection for demanding work environments. Designed for all-day comfort, it features a 4-point ratchet suspension and is constructed from durable High-Density Polyethylene (HDPE). This non-vented, short-brim hard hat is suitable for various industrial and construction applications.
  • Provides Class C, G, and E head protection.
  • Features a 4-point ratchet suspension for a secure, universal fit.
  • Constructed from High-Density Polyethylene (HDPE) for durability.
  • Non-vented design.
  • Equipped with accessory slots for compatibility with headgear.
  • Meets ANSI Type I standards.
  • Resistant to temperatures from -30°C to +50°C.
  • Certified to ANSI/ISEA Z89.1 and CSA Z94.1-15 standards.
Catalog No. 50-005-09045 Supplier MSC Supplier No. 16795676
This industrial hard hat provides certified head protection for demanding work environments. Designed for all-day comfort, it features a 4-point ratchet suspension and is constructed from durable High-Density Polyethylene (HDPE). This non-vented, short-brim hard hat is suitable for various industrial and construction applications.
  • Provides Class C, G, and E head protection.
  • Features a 4-point ratchet suspension for a secure, universal fit.
  • Constructed from High-Density Polyethylene (HDPE) for durability.
  • Non-vented design.
  • Equipped with accessory slots for compatibility with headgear.
  • Meets ANSI Type I standards.
  • Resistant to temperatures from -30°C to +50°C.
  • Certified to ANSI/ISEA Z89.1 and CSA Z94.1-15 standards.