Learn More
Honeywell™ NorthFlex Light Task Plus 3™ Dyneema™ Cut-Resistant Gloves, Polyurethane Coated
Description
Suited for material handling and assembly work involving sharp objects like sheet metal, metal shavings, glass and metal parts—anywhere mechanical hazards exist
- Seamless Dyneema liner is up to 15 times stronger than steel (weight for weight) and outperforms aramid fibers in cut and abrasion resistance
- Dyneema shell is stretchy and form fitting, allowing dexterity
- PU coating provides abrasion resistance, but is light enough to ensure workers retain dexterity and tactility
- Gray color hides dirt, making it a good choice for many tough and dirty applications
- Uncoated back of glove assures ventilation and comfort
- Contains no reaction causing, natural rubber proteins
- Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Specifications
| Construction | Seamless Knit |
| Color | Gray, White |
| Cut Resistance Rating | ANSI A1 |
| Material | Dyneema |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| EN388 Standard | EN 388 |
| For Use With (Application) | sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work |
| Glove Cuff Style | Knit |
| Model | NFD16G/7S |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.