Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus 3™ Dyneema™ Cut-Resistant Gloves, Polyurethane Coated

Catalog No. 19130271B
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-271B Medium
19-130-271A Small
19-130-271C Large
19-130-271D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-271B Supplier Honeywell™ Supplier No. NFD16G/8M
This item is not returnable. View return policy

Medium-weight cut-resistant glove that offers a great compromise between mechanical protection and wearer comfort.

Suited for material handling and assembly work involving sharp objects like sheet metal, metal shavings, glass and metal parts—anywhere mechanical hazards exist

  • Seamless Dyneema liner is up to 15 times stronger than steel (weight for weight) and outperforms aramid fibers in cut and abrasion resistance
  • Dyneema shell is stretchy and form fitting, allowing dexterity
  • PU coating provides abrasion resistance, but is light enough to ensure workers retain dexterity and tactility
  • Gray color hides dirt, making it a good choice for many tough and dirty applications
  • Uncoated back of glove assures ventilation and comfort
  • Contains no reaction causing, natural rubber proteins
  • Cuff over-stitching is color-coded by size to aid sorting after laundering

TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Color Gray, White
Cut Resistance Rating ANSI A1
Material Dyneema
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388
For Use With (Application) sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work
Glove Cuff Style Knit
Model NFD16G/8M
Series NorthFlex Light Task Plus 3™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.