Learn More
Honeywell™ North™ Lightning™ Series Safety Eyewear Replacement Lenses
Catalog No. 191300767
Change view
Click to view available options
Lens Tint:
Clear
Smoke
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-130-0767 Smoke
19-130-0766 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-0767 Supplier Honeywell™ Supplier No. 470RL
This item is not returnable. View return policy
Frameless Lightning Plus safety glasses offer full-view unilens construction. Interchangable lenses attach to temples that provide a secure fit. Prescription insert simplifies your safety program, allowing use by your entire workforce.
  • Anti-scratch, anti-UV 99.9% and anti-static lenses
  • Available in clear, smoke or mirrored tints
  • Meets ANSI Z87.1-2010 and tested to meet the requirements of the CSA Z94.3 standard
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint Smoke
Product Type Replacement lens