Learn More
Honeywell™ North™ Lightning™ T6500 Series Safety Eyewear
Catalog No. 191300746
Change view
Click to view available options
Lens Tint:
Clear
Temple Style:
Self-adjusting
Catalog No. Lens Tint Temple Style
19-130-0746 Clear Self-adjusting
1 options
This item is not returnable. View return policy
Catalog No. 19-130-0746 Supplier Honeywell™ Supplier No. T65005B
This item is not returnable. View return policy
Engineered for safety officers but designed for users
  • High base panoramic lens for a wide unobstructed field of view
  • Frame made of new Elasto LT chemical resistant material
  • New Soft-Touch comfort pads
  • Self-adjusting temples that insure a secure yet comfortable fit
  • The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
  • Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
  • Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-static, Scratch-resistant, UV-treated
Lens Tint Clear
Frame Color Black
Temple Style Self-adjusting