Learn More
Honeywell™ North™ Lightning™ T6550 Series Safety Eyewear
Catalog No. 191300765
Change view
Click to view available options
Frame Color:
Blue
Bronze
Lens Tint:
Amber
Mirror
Smoke
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-0765 Bronze Mirror
19-130-0757 Blue Mirror
19-130-0764 Bronze Amber
19-130-0763 Bronze Smoke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-0765 Supplier Honeywell™ Supplier No. T65505BRM
This item is not returnable. View return policy
Engineered for safety officers but designed for users
  • High base panoramic lens for a wide unobstructed field of view
  • Bowed temples designed to fit virtually anyone
  • New Soft-Touch comfort pads
  • Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
  • The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
  • Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-static, Scratch-resistant, UV-treated
Lens Tint Mirror
Frame Color Bronze
Temple Style Bowed w/soft-touch pads
Lens Material Polycarbonate