Learn More
Honeywell™ North™ Model 7190 N99 Air-Purifying Respirator
Catalog No. 19034625
Change view
Click to view available options
Product Type:
Air Purifying Respirator
Certifications/Compliance:
OSHA, NIOSH 42CFR84, ISO 9001
Catalog No. Product Type Certifications/Compliance
19-034-625 Air Purifying Respirator OSHA, NIOSH 42CFR84, ISO 9001
1 options
Catalog No. 19-034-625 Supplier Honeywell™ Supplier No. 7190N99
Economical and reusable
  • Non-oil particulate Model 7190 N99 provides a cool, comfortable alternative to disposable respirators
  • Coaxial valving system provides separate intake and outflow channels
  • Larger filter surface area offers low breathing resistance
  • Low-profile design fits easily under a welding helmet with no obstruction of vision
  • Replaceable N99 filter provides protection against solid particulates and non–oil-based aerosols
Compliance
Respirator meets OSHA requirements and is approved to NIOSH 42CFR84 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance OSHA, NIOSH 42CFR84, ISO 9001
Design Type Low profile
Disposable Single-use
Filter Type N99
Product Type Air Purifying Respirator
Size One Size
For Use With (Application) Provides protection against solid particulates and non-oil based aerosols
Includes (1 ) Respirator, (1) N99 Filter
Quantity Each