Learn More
Honeywell™ North™ N-Vision™ Safety Glasses
Description
Available in a wide range of lens tints, utilizing the exclusive 4A coating. Presscription insert available. Replaceable nose piece.
Specifications
Specifications
| Lens Coating | Anti-fog, Anti-static, Scratch-resistant |
| Lens Tint | Clear |
| Frame Color | Blue/Gray |
| No. per Pack | 10/Pk. |
| Temple Style | Non-Adjustable |
| Temple Color | Blue |
| Lens Material | Polycarbonate |
| Frame Material | Elasto LT Chemical Resistant Material |
| Frame Type | Wraparound |