Learn More
Honeywell™ North™ National Brand Antacid Tablets
Specifications
Specifications
| Product Type | Tums Antacid Tablet |
| Active Ingredients | Antacid |
| Format | Tablet |
| Flavor | Peppermint |
| For Use With (Application) | Fast relief from the symptoms of Acid indigestion, heartburn, gas and sour stomach |
| Quantity | 150 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.