Learn More
MSC North Nitrile Chemical Gloves, X-Large, 11.0 mil, Unsupported, 1 Pair
Catalog No. 5000506721
Encompass
These chemical resistant gloves are designed for tasks requiring dexterity and involving light-to-medium chemical applications. The unsupported design is economical and allows use with or without a knitted or woven liner.
  • X-Large size (numeric size 10)
  • Green color
  • Thickness of 11.0 mil
  • Unsupported
  • ANSI/ISEA abrasion resistance level 4
  • ANSI puncture level 1
  • Unlined
  • FDA approved
  • Overall length of 13.0 in
Catalog No. 50-005-06721 Supplier MSC Supplier No. 76807205
These chemical resistant gloves are designed for tasks requiring dexterity and involving light-to-medium chemical applications. The unsupported design is economical and allows use with or without a knitted or woven liner.
  • X-Large size (numeric size 10)
  • Green color
  • Thickness of 11.0 mil
  • Unsupported
  • ANSI/ISEA abrasion resistance level 4
  • ANSI puncture level 1
  • Unlined
  • FDA approved
  • Overall length of 13.0 in