Learn More
Honeywell™ NorthFlex Oil-Grip™ Nitrile Coated Nylon Gloves
Catalog No. 19045567
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
19-045-567 2X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-045-567 Supplier Honeywell™ Supplier No. NF35/11XXL
This item is not returnable. View return policy
Lightweight, 13-gauge nylon shell is comfortable and flexible, while the MicroFinish nitrile coating enhances grip capabilities in oily applications.
  • Nitrile coating is textured with thousands of microscopic pockets that disperse fluids away from the glove's surface
  • MicroFinish™ surface allows for a high level of surface contact between glove and oil saturated object
  • Two stage coating is impermeable to liquids, so that hands remain dry
  • Lightweight, 13-gauge seamless nylon liner is soft and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Dark colors hide dirt and grime for extended wear
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Nitrile
Color Green
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Glove Cuff Style Knit
Series NF35
Yarn Gauge 13