Learn More
Honeywell™ North™ O/H Pak First Aid Cabinets
Catalog No. 19065214
Change view
Click to view available options
Dimensions (L x W x H):
15 x 10.5 x 4.5 in.
Catalog No. Dimensions (L x W x H)
19-065214 15 x 10.5 x 4.5 in.
1 options
Catalog No. 19-065214 Supplier Honeywell™ Supplier No. 0190893462A/DEL
Easily wall mounted to provide a centralized location for medical supplies
Filled kits contain an assortment of first aid and medical supplies necessary to treat most minor injuries and pain.
  • Adhesive strips come in a variety of sizes and styles, including fingertip and knuckle bandages
  • Plastic and cloth bandages have a soft nonstick pad and are individually packaged in sterile wraps
  • Medicinals are packed in unit doses of two tablets per packet for convenient dispensing
  • Each unit dose pouch is color coded for quick identification; pouch is marked with expiration date
Order Info
Recommended kit size for number of people: Medium for 50-75, Large for 75-100, Deluxe for 100-125. Cabinets also available empty, so you can custom-stock your own first aid kit.
TRUSTED_SUSTAINABILITY
Specifications
Includes 1 in. x 2.5yd. Adhesive tape, Alcohol prep pads, ammonia inhalants, antibiotic ointment pouches, 3 oz. Antiseptic spray, 5 oz. Burn spray, 0.75 x 3 in. cloth adhesive bandage, 1 x 3 in. cloth adhesive bandage, cloth fingertip bandage, cloth knuckle bandage, cold pak, instant, eye dressing packet, 4 oz. eye-lert™ buffered eyewash, first aid guide, forceps, pointed, 2 in. offset™ bandage, pain-a-rest™ nonaspirin, pvp iodine wipes, 4 in. scissors, blunt, 3 x 3 in. sterile pads, triangular bandage, sterile, 0.5, 0.37, 0.87 in. x 5 yd. tri-cut tape
Dimensions (L x W x H) 15 x 10.5 x 4.5 in.