Learn More
Honeywell™ North™ Panther Hip-Pac Respirator
Catalog No. 19034613
Change view
Click to view available options
Duration:
10 min.
15 min.
Includes:
3000 psi aluminum cylinder, silicone twenty plus facepiece, regulator, medium, black, medium nose cup, silicone headstrap
Facepiece, regulator and 3000 psig hoop wrapped cylinder with quick-disconnect, silicone mask, polycarbonate lens
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration Includes
19-034-613 15 min. Facepiece, regulator and 3000 psig hoop wrapped cylinder with quick-disconnect, silicone mask, polycarbonate lens
18-244 10 min. 3000 psi aluminum cylinder, silicone twenty plus facepiece, regulator, medium, black, medium nose cup, silicone headstrap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-613 Supplier Honeywell™ Supplier No. P968500
This product provides air from a remote air source for entry into and escape from IDLH atmospheres. The escape cylinder is available in 5-, 10- and 15-minute durations.
The Pressure Demand SAR with Escape Cylinder (Hip-Pac) offers the same reliable facepieces and second stage regulators as the Cougar and 1997-style Panther SCBAs. It allows the user to comfortably wear the respirator to enter an IDLH environment while breathing from a remote air source. Should the air supply be interrupted, the user can switch to the cylinder for escape.
All airline respirators must be used with NIOSH-approved lightweight or high-performance airline hose for use up to 300 ft. (91m) from air supply.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Product Type Standard Escape Respirator
Duration 15 min.
Cycle Time 15 min.
For Use With (Application) Chemical, construction, food services, manufacturing, military, mining, oil and gas, pharmaceutical
Includes Facepiece, regulator and 3000 psig hoop wrapped cylinder with quick-disconnect, silicone mask, polycarbonate lens
Weight (Metric) 6.8 kg
Size Medium
Weight (English) 15 lb.