Learn More
Honeywell™ North™ Premier 611 Half-Mask Respirators for Paints and Pesticides
Catalog No. 176315
Change view
Click to view available options
Size:
Medium
Large
Includes:
Cartridges and Filter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
17-631-5 Medium -
17-631-3 Large Cartridges and Filter
17-631-6 Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-631-5 Supplier Honeywell™ Supplier No. 312120
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Silicone half-mask respirators can be used with N95 or P95 cartridges and filters depending on specific environment.
  • Complies with NIOSH 42 CFR 84 standards
  • Half-mask design same as Series 6100 respirator
  • Easy-to-clean silicone
  • Dual-cartridge features a soft sealing flange and pliable nosepiece for maximum comfort
  • Nonslip cradle suspension and floating yoke ensure a secure fit
  • Additional room for protective eyewear
  • P95 filter for paints and pesticides containing oil; the N95 filter is suitable for non-oil environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications (NIOSH) NIOSH 42 CFR Part 84
Material Silicone
Product Type Half-Mask Respirator
Size Medium
For Use With (Application) Chemical, construction, food services, forestry, government, manufacturing, military, mining, municipal services, oil and gas, pharmaceutical, steel and metals, welding
For Use With (Equipment) T Series Cartridge
Model 312120