Learn More
MSC North Safety Glasses, Polycarbonate, Gray, Scratch-Resistant, ANSI Z87.1, 1 ea
Catalog No. 5000506927
Encompass
These safety glasses feature scratch-resistant polycarbonate gray lenses and a half-framed style for reliable eye protection. They offer a wrap-around design with a gray plastic frame and non-adjustable temples, suitable for various work environments.
  • Tinted style
  • Scratch-resistant lens coating
  • Half-framed, wrap-around design
  • Gray polycarbonate lenses
  • Gray plastic frame and temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Padded temple tips
Catalog No. 50-005-06927 Supplier MSC Supplier No. 72472798
These safety glasses feature scratch-resistant polycarbonate gray lenses and a half-framed style for reliable eye protection. They offer a wrap-around design with a gray plastic frame and non-adjustable temples, suitable for various work environments.
  • Tinted style
  • Scratch-resistant lens coating
  • Half-framed, wrap-around design
  • Gray polycarbonate lenses
  • Gray plastic frame and temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Padded temple tips