Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Sparkguard Headgear

Catalog No. 191303961
Change view
Click to view available options
Large Full Floating Mount Sparkguard Headgear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-3961 Large Full Floating Mount Sparkguard Headgear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3961 Supplier Honeywell™ Supplier No. KHG5001

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Basic, simple-to-use headgear for all applications. A protected mounting system helps eliminate fluids from entering the backside of the faceshield, while an unprotected mounting system offers quick visor replacement. Available with the SmoothLok ratchet and Protected Fit systems.

Patented non-slip ratchet system is easy to use and offers precise size adjustments. The SmoothLok ratchet is available on the KHG Series headgear for quick, easy and perfect fit. The 5000 Series comes with Protected Fit holding system for faceshield to help reinforce splash protection to inside of the faceshield.

  • Accepts a wide range of faceshield styles
  • Large sparkguard provides excellent protection
  • Cross strap adjusts to allow an exceptional fit
  • Full floating mount sparkguard provides comfortable wear even with heavy faceshield and continuous use
  • Meets ANSI Z87.1-2010
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Faceshield Visor
Includes SmoothLok ratchet adjustment
Material Plastic
Product Type Headgear
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.