Learn More
Honeywell™ North™ Food Service Burn First Aid Kits
Specifications
Specifications
| For Use With (Application) | For treating burn accidents that may occur in kitchens or other food processing operations |
| Product Type | 10-Unit First Aid Kit |
| Includes | 10-units; 5 water-jel dressings, self-adherent wrap, pair medical gloves, scissors, essential burn kit items for kitchens or other food processing operations |
| Quantity | 10 Pk. |