Learn More
Honeywell™ North™ Specialized First Aid Kits
Catalog No. 19088243
Change view
Click to view available options
Product Type:
Pocket First Aid Kit
Catalog No. Product Type
19-088243 Pocket First Aid Kit
1 options
Catalog No. 19-088243 Supplier Honeywell™ Supplier No. 123980
Utility Burn Kit This kit is designed for utilities and provides the essential items to quickly treat burn emergencies.
Kit Contents: Medical gloves (pair), Scissors (angled, red handle), Water-Jel™ Dressing (4 x 4 in.), 2 Water-Jel™ Dressing (4 x 16 in.), Water-Jel™ Face Mask (12 x 16 in.)
Order Info
Unit fill packs are easy to access and easy to re-order.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pocket First Aid Kit