Learn More
MSC Honeywell North Chemical-Resistant Gloves, Butyl, 32 mil, Unsupported, 1 ea
Catalog No. 5000510001
Encompass
These unsupported butyl gloves provide exceptional protection against a broad range of toxic chemicals, including ketones and esters. Designed with a 32 mil thickness, they offer the highest permeation resistance to gases and water vapors, making them ideal for hazardous material handling and chemical processing environments. The multi-dip manufacturing process ensures superior quality by creating multiple polymer layers for enhanced durability.
  • Provides high chemical resistance to ketones like MEK, MIBK, and acetone
  • Offers superior protection against esters such as tricresyl phosphate and ethyl acetate
  • Features a 14 in. length and rolled, beaded cuff for extended protection
  • Constructed from durable, unlined butyl for high-performance safety applications
  • Includes a rough grip pattern for improved dexterity and handling
Catalog No. 50-005-10001 Supplier MSC Supplier No. 01904192
These unsupported butyl gloves provide exceptional protection against a broad range of toxic chemicals, including ketones and esters. Designed with a 32 mil thickness, they offer the highest permeation resistance to gases and water vapors, making them ideal for hazardous material handling and chemical processing environments. The multi-dip manufacturing process ensures superior quality by creating multiple polymer layers for enhanced durability.
  • Provides high chemical resistance to ketones like MEK, MIBK, and acetone
  • Offers superior protection against esters such as tricresyl phosphate and ethyl acetate
  • Features a 14 in. length and rolled, beaded cuff for extended protection
  • Constructed from durable, unlined butyl for high-performance safety applications
  • Includes a rough grip pattern for improved dexterity and handling