Learn More
Honeywell™ North™ Three-Piece Rainsuits
Catalog No. 19310025
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-310-025 Medium
19-310-024 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-310-025 Supplier Honeywell™ Supplier No. 85JPHM
This item is not returnable. View return policy
Suits include bib pants, jacket and hood
  • Resistant to abrasions and tearing
  • Flame retardant
  • PVC
  • Color: Yellow
  • 35 mil style has polyester backing
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Polyester/PVC
Size Medium