Learn More
Honeywell™ North™ Tornado Safety Glasses
Catalog No. 17986701
Change view
Click to view available options
Frame Color:
Black, Red
Catalog No. Frame Color
17-986-701 Black, Red
1 options
This item is not returnable. View return policy
Catalog No. 17-986-701 Supplier Honeywell™ Supplier No. T57505B
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Great coverage provided by the Tornado F5 is offered in 3 frame colors and two temple styles, providing terrific fit and style options.
Replaceable lenses are offered in the North exclusive 4A coating. Meets ANSI Z87.1-2010 and is tested by Honeywell to meet the requirements of the CSA Z94.3 standard.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant, UV-treated
Lens Tint Clear
Frame Color Black, Red
No. per Pack 12/Pk.