Learn More
Honeywell™ North™ Unisorb Cartridges
Catalog No. 19003438
Change view
Click to view available options
Filter Type:
N95
Unisorb Multi-Contaminant Cartridge
Quantity:
24/Pk.
6/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Quantity
19-003-438 N95 24/Pk.
18-999-1306 Unisorb Multi-Contaminant Cartridge 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-003-438 Supplier Honeywell™ Supplier No. T106010
Eliminate the need to stock contaminant-specific cartridges to simplify training, ordering, and inventory. Survivair T-Series Twist-Lock Cartridges provide an audible click indicating a positive lock on connector. NIOSH approved for up to 16 contaminants.
These economical cartridges offer lower breathing resistance than many similar products and feature a low profile for better visibility and weight distribution. An audible “locking” bayonet connection confirms cartridge attachment, preventing cross-threading and under/over tightening concerns. And for maximum flexibility, both standard and HEPA cartridges can be used on any Willson ValuAir™ Plus, Premier™ 6800, 6100 half-mask, or 6400/6500 Series full-facepiece respirators.
Used alone, the standard Unisorb cartridge offers protection against a variety of contaminants, including hydrogen fluoride. When paired with a Willson N95 or P95 prefilter and filter retainer (P95 filter sold separately), the Unisorb is approved for use in environments containing dusts, mists, paint spray and pesticides. The Unisorb cartridge with P100 filter does not require a prefilter and also offers NIOSH-approved protection against asbestos-containing dusts and mists.
  1. Standard Unisorb cartridge requires a Willson N95 or P95 prefilter.
  2. Unisorb cartridge with P100 filter only.
  3. Dusts, mists, fumes have a TWA >0.05mg/m.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Filter Type N95
Product Type Filter
For Use With (Equipment) Respirator
Quantity 24/Pk.