Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NorthernLights Streptavidin NL557

Catalog No. NL999
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL999 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL999 Supplier R&D Systems™ Supplier No. NL999
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry

  • Maximum absorption 557nm; Maximum emission 574nm
  • Phosphate-buffered saline (PBS) containing <0.1% v/v sodium azide
  • Provided as a liquid

Applications:

Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Quantity 0.5 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.