Learn More
MSC North NorthFlex Cut-Resistant Gloves, ANSI A4, Palm-Coated, 10/X-Large, 1 Pair
Catalog No. 5000508942
Encompass
These cut-resistant gloves are engineered with a lightweight 13 gauge fiber blend designed to provide reliable hand protection in hazardous environments. They feature a textured foamed nitrile palm coating that enhances grip, making them suitable for metalworking and general maintenance tasks where cut resistance is required.
  • Proprietary cut-resistant fiber knit blend for optimal dexterity
  • Textured foamed nitrile palm dip for improved grip
  • Blended fibers provide extended wear and enhanced protection
  • Color-coded cuff over-edge stitching for efficient size sorting after laundering
  • ANSI/ISEA cut resistance level A4
  • Withstands commercial laundry detergents up to pH 14 and temperatures up to 49°C (120°F)
  • Warning: gloves are cut-resistant, not cut-proof
Catalog No. 50-005-08942 Supplier MSC Supplier No. 56155849
Add to Cart
Add to Cart
These cut-resistant gloves are engineered with a lightweight 13 gauge fiber blend designed to provide reliable hand protection in hazardous environments. They feature a textured foamed nitrile palm coating that enhances grip, making them suitable for metalworking and general maintenance tasks where cut resistance is required.
  • Proprietary cut-resistant fiber knit blend for optimal dexterity
  • Textured foamed nitrile palm dip for improved grip
  • Blended fibers provide extended wear and enhanced protection
  • Color-coded cuff over-edge stitching for efficient size sorting after laundering
  • ANSI/ISEA cut resistance level A4
  • Withstands commercial laundry detergents up to pH 14 and temperatures up to 49°C (120°F)
  • Warning: gloves are cut-resistant, not cut-proof