Learn More
MSC North Safety NorthFlex Cut Resistant Sleeve, 10 Gauge, 16 in., 1/ea.
Catalog No. 5000508156
Encompass
This cut-resistant sleeve provides advanced protection against slash, abrasion, and cut hazards in industrial and food service environments. Engineered with a 10-gauge Spectra® fiber and glass fiber blend, the 16 in. sleeve offers high-performance durability and low-linting properties to maintain cleanliness during use. Featuring a knit wrist design, it ensures a comfortable fit while maintaining manual dexterity for specialized tasks.
  • Provides excellent protection from cut, slash, and abrasion hazards
  • Constructed from Spectra® fiber, which is 10X stronger than steel by weight
  • Designed with low-linting materials suitable for food service applications
  • ANSI/ISEA cut level 5 and EN 388 cut level 5 rating
  • ANSI/ISEA abrasion level 4
  • Features a comfortable knit wrist closure
Catalog No. 50-005-08156 Supplier MSC Supplier No. 00774893
This cut-resistant sleeve provides advanced protection against slash, abrasion, and cut hazards in industrial and food service environments. Engineered with a 10-gauge Spectra® fiber and glass fiber blend, the 16 in. sleeve offers high-performance durability and low-linting properties to maintain cleanliness during use. Featuring a knit wrist design, it ensures a comfortable fit while maintaining manual dexterity for specialized tasks.
  • Provides excellent protection from cut, slash, and abrasion hazards
  • Constructed from Spectra® fiber, which is 10X stronger than steel by weight
  • Designed with low-linting materials suitable for food service applications
  • ANSI/ISEA cut level 5 and EN 388 cut level 5 rating
  • ANSI/ISEA abrasion level 4
  • Features a comfortable knit wrist closure