These safety glasses feature injection-molded lenses and temples, both made from 100% polycarbonate, ensuring impact resistance and durability. Designed for eye hazard areas, they offer optically correct 180° peripheral vision and absorb over 99.9% of harmful UV light. Vented sideshields facilitate airflow, reducing heat buildup and preventing lens fogging, while providing crucial side protection.
- Injection-molded lenses and temples
- Constructed from 100% polycarbonate for impact resistance
- Vented sideshields for airflow and to reduce heat buildup and lens fogging
- Provides side protection against impact
- Polycarbonate lens absorbs more than 99.9% of harmful UV light
- Optically correct 180° peripheral vision
- Universal-fit nose bridge and integrally molded browguard
- Meets ANSI Z87.1 standards for impact and particulate protection