Learn More
TARGETMOL CHEMICALS INC Nortriptyline 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7384425MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Nortriptyline is an antidepressant compound that enhances corticosteroid sensitivity of blood T-cells in patients with chronic obstructive pulmonary disease (COPD) and may be used for smoking cessation. Purity 99.89%
Catalog No. 50-287-6562