Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nortriptyline | 72-69-5 | 99.9% | 263.38 | C19H21N | 5 MG
SDP

Catalog No. 5000228961
Encompass_Preferred

Nortriptyline is a tricyclic antidepressant used as a research chemical and supplied as a high-purity solid for laboratory applications. It is provided in small pack sizes with defined storage conditions to preserve stability.

  • High reported purity (99.9%).
  • White to off-white solid, suitable for analytical and biological research.
  • Molecular weight 263.38 g/mol; CAS number 72-69-5.
  • Available in small research pack sizes, including 5 mg.
  • Store at 4°C and protect from light; in solvent, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-28961 Supplier Medchemexpress LLC Supplier No. HY1186205MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nortriptyline is a tricyclic antidepressant used as a research chemical and supplied as a high-purity solid for laboratory applications. It is provided in small pack sizes with defined storage conditions to preserve stability.

  • High reported purity (99.9%).
  • White to off-white solid, suitable for analytical and biological research.
  • Molecular weight 263.38 g/mol; CAS number 72-69-5.
  • Available in small research pack sizes, including 5 mg.
  • Store at 4°C and protect from light; in solvent, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.