Learn More
Medchemexpress LLC Nortriptyline-d3 hydrochloride | 203784-52-5 | 99.7% | 1 MG
SDP
Catalog No. 5000413004
Encompass_Preferred
Nortriptyline-d3 (hydrochloride) is the deuterium labeled Nortriptyline hydrochloride. It is a tricyclic antidepressant and the main active metabolite of Amitriptyline, used to relieve the symptoms of depression. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has garnered attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13004 Supplier Medchemexpress LLC Supplier No. HYB1417S1MG
Nortriptyline-d3 (hydrochloride) is the deuterium labeled Nortriptyline hydrochloride. It is a tricyclic antidepressant and the main active metabolite of Amitriptyline, used to relieve the symptoms of depression. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has garnered attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.