Learn More
Medchemexpress LLC Nortriptyline-d3 hydrochloride | 203784-52-5 | MFCD00198172 | 99.7% | 302.86 g/mol | C19H19D3ClN | 10 MG
SDP
Catalog No. 5000298036
Encompass_Preferred
Nortriptyline-d3 hydrochloride is a deuterium-labeled internal standard of nortriptyline (desmethylamitriptyline) intended for analytical and research use. It is provided as a white to off-white solid with high isotopic enrichment and suitability for quantitative LC-MS/MS assays, method development, and pharmacokinetic studies.
  • Stable-isotope labeled internal standard for mass spectrometry.
  • High purity suitable for quantitative analysis.
  • Provided as a solid convenient for storage and handling.
  • Available in multiple small package sizes for analytical workflows.
  • Suitable for method development, validation, and pharmacokinetic studies.
Catalog No. 50-002-98036 Supplier Medchemexpress LLC Supplier No. HYB1417S10MG
Nortriptyline-d3 hydrochloride is a deuterium-labeled internal standard of nortriptyline (desmethylamitriptyline) intended for analytical and research use. It is provided as a white to off-white solid with high isotopic enrichment and suitability for quantitative LC-MS/MS assays, method development, and pharmacokinetic studies.
  • Stable-isotope labeled internal standard for mass spectrometry.
  • High purity suitable for quantitative analysis.
  • Provided as a solid convenient for storage and handling.
  • Available in multiple small package sizes for analytical workflows.
  • Suitable for method development, validation, and pharmacokinetic studies.