Learn More
TARGETMOL CHEMICALS INC Nortriptyline hydrochloride
SDP
Catalog No. 502879609
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Nortriptyline hydrochloride (ELF-101 hydrochloride) is a metabolite of AMITRIPTYLINE that is also used as an antidepressive agent. Nortriptyline is used in major depression, dysthymia, and atypical depressions. Purity 99.71%
Catalog No. 50-287-9609 Supplier TARGETMOL CHEMICALS INC Supplier No. T1327500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Nortriptyline hydrochloride (ELF-101 hydrochloride) is a metabolite of AMITRIPTYLINE that is also used as an antidepressive agent. Nortriptyline is used in major depression, dysthymia, and atypical depressions. Purity 99.71%