Learn More
Selleck Chemical LLC Nortriptyline hydrochloride S3698-25mg
SDP
Supplier:  Selleck Chemical LLC S369825MG
Encompass_Preferred
Nortriptyline hydrochloride (Desitriptyline ELF-101 EN-7048 Desmethylamitriptyline) is the hydrochloride salt form of nortriptyline a tricyclic antidepressant agent used for short-term treatment of various forms of depression
Catalog No. 50-310-8785