Learn More
Apexbio Technology LLC Nortriptyline (hydrochloride) 894-71-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088760
Encompass_Preferred
Nortriptyline hydrochloride (CAS 894-71-3) is a dibenzocycloheptene-derived tricyclic antidepressant that inhibits monoamine reuptake transporters It selectively blocks the norepinephrine transporter (NET) with a Ki of 3 4 nM and inhibits the serotonin transporter (SERT) with a Ki of 161 nM thereby increasing synaptic concentrations of these neurotransmitters in the central nervous system In vitro nortriptyline dose-dependently suppresses spontaneous migration and chemotaxis of human polymorphonuclear cells In vivo studies in rodents indicate partial blockade of noradrenaline depletion in nerve terminals at higher doses Nortriptyline is frequently employed in research on monoaminergic neurotransmission and antidepressant pharmacology
Catalog No. 50-000-88760 Supplier Apexbio Technology LLC Supplier No. C40775.1
Add to Cart
Add to Cart
Nortriptyline hydrochloride (CAS 894-71-3) is a dibenzocycloheptene-derived tricyclic antidepressant that inhibits monoamine reuptake transporters It selectively blocks the norepinephrine transporter (NET) with a Ki of 3 4 nM and inhibits the serotonin transporter (SERT) with a Ki of 161 nM thereby increasing synaptic concentrations of these neurotransmitters in the central nervous system In vitro nortriptyline dose-dependently suppresses spontaneous migration and chemotaxis of human polymorphonuclear cells In vivo studies in rodents indicate partial blockade of noradrenaline depletion in nerve terminals at higher doses Nortriptyline is frequently employed in research on monoaminergic neurotransmission and antidepressant pharmacology