Learn More
Medchemexpress LLC Nortriptyline hydrochloride | 894-71-3 | 99.98% | 1 G
SDP
Catalog No. 5000413405
Encompass_Preferred
Nortriptyline (Desmethylamitriptyline) hydrochloride is the main active metabolite of Amitriptyline and acts as a tricyclic antidepressant. It is also a potent autophagy inhibitor and has anticancer effects.
  • Main active metabolite of Amitriptyline
  • Tricyclic antidepressant
  • Potent autophagy inhibitor
  • Has anticancer effects
Catalog No. 50-004-13405 Supplier Medchemexpress LLC Supplier No. HYB14171G
Nortriptyline (Desmethylamitriptyline) hydrochloride is the main active metabolite of Amitriptyline and acts as a tricyclic antidepressant. It is also a potent autophagy inhibitor and has anticancer effects.
  • Main active metabolite of Amitriptyline
  • Tricyclic antidepressant
  • Potent autophagy inhibitor
  • Has anticancer effects