Learn More
Medchemexpress LLC Nortriptyline hydrochloride | 894-71-3 | 99.98% | 1 ML
SDP
Catalog No. 5000413436
Encompass_Preferred
Nortriptyline hydrochloride (Desmethylamitriptyline) is the main active metabolite of Amitriptyline, functioning as a tricyclic antidepressant. It also acts as a potent autophagy inhibitor and demonstrates anticancer effects.
  • Potent autophagy inhibitor
  • Anticancer effects
  • Inhibits Norepinephrine transporter (NET, SLC6A2)
  • Blocks Norepinephrine reuptake more potently than Serotonin
  • Reduces viability and induces apoptosis in bladder cancer cells
  • Inhibits bladder tumor growth in vivo
Catalog No. 50-004-13436 Supplier Medchemexpress LLC Supplier No. HYB141710MM/1ML
Nortriptyline hydrochloride (Desmethylamitriptyline) is the main active metabolite of Amitriptyline, functioning as a tricyclic antidepressant. It also acts as a potent autophagy inhibitor and demonstrates anticancer effects.
  • Potent autophagy inhibitor
  • Anticancer effects
  • Inhibits Norepinephrine transporter (NET, SLC6A2)
  • Blocks Norepinephrine reuptake more potently than Serotonin
  • Reduces viability and induces apoptosis in bladder cancer cells
  • Inhibits bladder tumor growth in vivo