Learn More
Medchemexpress LLC Nortriptyline hydrochloride | 894-71-3 | 99.98% | 100 MG
SDP
Catalog No. 5000413403
Encompass_Preferred
Nortriptyline hydrochloride is the main active metabolite of Amitriptyline, classified as a tricyclic antidepressant. It functions as a potent autophagy inhibitor and demonstrates anticancer effects, intended for research use only.
  • Inhibits the Norepinephrine transporter (NET, SLC6A2)
  • Reduces viability of bladder cancer cells
  • Induces cell cycle arrest
  • Induces apoptosis in bladder cancer cells
  • Blocks reuptake of Norepinephrine more potently than Serotonin
Catalog No. 50-004-13403 Supplier Medchemexpress LLC Supplier No. HYB1417100MG
Nortriptyline hydrochloride is the main active metabolite of Amitriptyline, classified as a tricyclic antidepressant. It functions as a potent autophagy inhibitor and demonstrates anticancer effects, intended for research use only.
  • Inhibits the Norepinephrine transporter (NET, SLC6A2)
  • Reduces viability of bladder cancer cells
  • Induces cell cycle arrest
  • Induces apoptosis in bladder cancer cells
  • Blocks reuptake of Norepinephrine more potently than Serotonin