Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nortriptyline Hydrochloride, MP Biomedicals™

Catalog No. ICN15355583 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15355583 250 mg
ICN15355501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15355583 Supplier MP Biomedicals, Inc Supplier No. 0215355583
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 894-71-3
Molecular Formula C19H22ClN
Molecular Weight (g/mol) 299.84
MDL Number MFCD00058024
InChI Key SHAYBENGXDALFF-UHFFFAOYSA-N
Synonym nortriptyline hydrochloride, pamelor, allegron, altilev, nortrilen, desmethylamitriptyline hydrochloride, acetexa, psychostyl, sensival, vividyl
PubChem CID 441358
IUPAC Name hydrogen methyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CNCCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Color White
Quantity 250 mg
Formula Weight 299.8
Physical Form Solid
Chemical Name or Material Nortriptyline Hydrochloride
EINECSNumber 212-973-0
storageNote1 Protect from light.
Recommended Storage Store at room temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.