Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nortriptyline Hydrochloride, USP, 97-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604852 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-852 500 g Amber Glass Bottle
18-604-850 5 g Amber Glass Bottle
18-604-851 25 g Amber Glass Bottle
18-604-853 100 g Amber Glass Bottle
18-604-854 1 kg Amber Glass Bottle
18-604-855 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-852 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1054500GMBL
Only null left
Add to Cart
Add to Cart

N1054, 894-71-3, C19H21N·HCl

Spectrum™ Chemical Nortriptyline Hydrochloride, USP is a tricyclic antidepressant used to treat nocturnal bedwetting. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 894-71-3
Molecular Formula C19H22ClN
Molecular Weight (g/mol) 299.84
MDL Number MFCD00058024
InChI Key SHAYBENGXDALFF-UHFFFAOYSA-N
IUPAC Name hydrogen methyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CNCCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 97 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.