Learn More
Cayman Chemical Nortriptyline (hydrochloride), Purity: ≥98%, CAS: 894-71-3, 100 mg
SDP
Catalog No. 502971036
Encompass_Preferred
A tricyclic antidepressant that blocks norepinephrine and serotonin transporters (KDs = 4.4 and 18 nM, respectively) more potently than the dopamine transporter (KD = 1.1 µM)
Catalog No. 50-297-1036 Supplier Cayman Chemical Supplier No. 15904100 MG
A tricyclic antidepressant that blocks norepinephrine and serotonin transporters (KDs = 4.4 and 18 nM, respectively) more potently than the dopamine transporter (KD = 1.1 µM)