Learn More
Medchemexpress LLC HY-W018601 100mg Medchemexpress, Nortropine CAS:538-09-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW018601100MG
Encompass_Preferred
Medchemexpress, HY-W018601 100mg Nortropine CAS:538-09-0 Nortropine (Nortropenol), isolated from the total alkaloids of Convolvulus subhirsutus, is an intermediate in tropine breakdown and reactions leading to succinate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3426