Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nortropine hydrochloride | 14383-51-8 | MFCD01711246 | 99.7% | 163.65 g/mol | C7H14ClNO | 1 ML
SDP

Catalog No. 5000210396
Encompass_Preferred

Nortropine hydrochloride is a tropane alkaloid intermediate used in organic synthesis and research. It is available as a solid and as a ready-to-use 10 mM solution in DMSO, and is characterized by CAS 14383-51-8, formula C7H14ClNO, and molecular weight 163.65 g/mol.

  • Synonym: nortropenol hydrochloride.
  • Available as a 10 mM, 1 mL solution in DMSO and as solid quantities (1 g, 5 g, 10 g).
  • Purity: 99.7%.
  • Storage: solid stored sealed at 4°C away from moisture; solution stability at -80°C (6 months) or -20°C (1 month).
  • Typical use: intermediate in synthesis and research involving tropane alkaloids.

Catalog No. 50-002-10396 Supplier Medchemexpress LLC Supplier No. HYW06691510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nortropine hydrochloride is a tropane alkaloid intermediate used in organic synthesis and research. It is available as a solid and as a ready-to-use 10 mM solution in DMSO, and is characterized by CAS 14383-51-8, formula C7H14ClNO, and molecular weight 163.65 g/mol.

  • Synonym: nortropenol hydrochloride.
  • Available as a 10 mM, 1 mL solution in DMSO and as solid quantities (1 g, 5 g, 10 g).
  • Purity: 99.7%.
  • Storage: solid stored sealed at 4°C away from moisture; solution stability at -80°C (6 months) or -20°C (1 month).
  • Typical use: intermediate in synthesis and research involving tropane alkaloids.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.