Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nortropine (nortropenol) | 538-09-0 | MFCD00047140 | 99.6% | 127.19 g/mol | C7H13NO | 10 G
SDP

Catalog No. 5000301064
Encompass_Preferred

Nortropine (nortropenol) is an alkaloid isolated from the total alkaloids of Convolvulus subhirsutus. It is used as a small-molecule research reagent and an intermediate in synthetic chemistry and metabolic studies involving tropine breakdown.

  • CAS number 538-09-0.
  • Molecular formula C7H13NO.
  • Molecular weight 127.19 g/mol.
  • Used as an intermediate in tropine metabolism and succinate-forming reactions.
  • Supplied at research-grade purity for laboratory synthesis and analysis.

Catalog No. 50-003-01064 Supplier Medchemexpress LLC Supplier No. HYW01860110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Nortropine (nortropenol) is an alkaloid isolated from the total alkaloids of Convolvulus subhirsutus. It is used as a small-molecule research reagent and an intermediate in synthetic chemistry and metabolic studies involving tropine breakdown.

  • CAS number 538-09-0.
  • Molecular formula C7H13NO.
  • Molecular weight 127.19 g/mol.
  • Used as an intermediate in tropine metabolism and succinate-forming reactions.
  • Supplied at research-grade purity for laboratory synthesis and analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.